missing translation for 'onlineSavingsMsg'
Learn More

p-Chloroaniline Hydrochloride, TRC

Product Code. 30357463
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357463 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357463 Supplier Toronto Research Chemicals Supplier No. TRCC3644501G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Chloroaniline hydrochloride
CAS 20265-96-7
Molecular Formula C6 H6 Cl N . Cl H
Quantity 1 g
Synonym Benzenamine, 4-chloro-, hydrochloride (1:1), Aniline, p-chloro-, hydrochloride (8CI), Benzenamine, 4-chloro-, hydrochloride (9CI), 4-Chloroaniline hydrochloride, 4-Chloroaniline monohydrochloride, 4-Chloroanilinium chloride, 4-Chlorobenzenamine hydrochloride, 4-Chlorophenylammonium chloride, p-Chloroaniline hydrochloride, p-Chloroanilinium chloride, p-Chlorophenylamine hydrochloride
InChI Formula InChI=1S/C6H6ClN.ClH/c7-5-1-3-6(8)4-2-5;/h1-4H,8H2;1H
SMILES Cl.Nc1ccc(Cl)cc1
IUPAC Name 4-chloroaniline;hydrochloride
Molecular Weight (g/mol) 164.03
Formula Weight 162.9956
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.