missing translation for 'onlineSavingsMsg'
Learn More

p-Coumaric Acid, MP Biomedicals

Product Code. 11423205 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423205 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423205 Supplier MP Biomedicals Supplier No. 0521390505
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7400-08-0
Molecular Formula C9H8O3
Quantity 5 g
Synonym p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid
InChI Key NGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILES C1=CC(=CC=C1C=CC(=O)O)O
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 164.16
PubChem CID 637542
ChEBI CHEBI:32374
Formula Weight 164.16
Chemical Name or Material p-Coumaric Acid
Show More Show Less
compliance-icons
Product Identifier
  • p-Coumaric Acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.