missing translation for 'onlineSavingsMsg'
Learn More

p-Coumaric acid, TRC

Product Code. 30364851
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364851 5 g -
30353257 100 g 100g
30354791 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364851 Supplier Toronto Research Chemicals Supplier No. TRCC7553655G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7400-08-0
Molecular Formula C9 H8 O3
Molecular Weight (g/mol) 164.16
Synonym 4-trans-4-Hydroxycinnamic acid
IUPAC Name 3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)C=Cc1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)
Formula Weight 164.0473
Percent Purity >95%
Chemical Name or Material Coumaric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.