Learn More
p-Cresol-d7, TRC
Product Code. 30368018
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368018 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30368018 Supplier Toronto Research Chemicals Supplier No. TRCC78190225MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material p-Cresol-d7
CAS 202325-52-8
Purity Grade Notes HPLC
Molecular Formula C7 2H7 H O
Quantity 25 mg
Synonym Phen-2,3,5,6-d4-ol, 4-(methyl-d3)- (9CI), 2,3,5,6-Tetradeuterio-4-(trideuteriomethyl)phenol, 4-(Methyl-d3)phen-2,3,5,6-d4-ol, p-Cresol-d7, 4-Methylphenol-D7
InChI Formula InChI=1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3/i1D3,2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1O)C([2H])([2H])[2H]
IUPAC Name 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenol
Molecular Weight (g/mol) 115.18
Formula Weight 115.1015
Percent Purity >95