missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxypropiophenone, TRC

Product Code. 30407172
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 30407172

Brand: Toronto Research Chemicals TRCH952515500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Hydroxypropiophenone
CAS 70-70-2
Molecular Formula C9H10O2
Quantity 500 g
Synonym 1-(4-Hydroxyphenyl)-1-propanone, 1-(p-Hydroxyphenyl)-1-propanone, 4-Propanoylphenol, 4-Propionylphenol, 4'-Hydroxypropiophenone, B 360, Bio-fren, Ethyl 4-hydroxyphenyl ketone, Ethyl p-hydroxyphenyl ketone, Frenantol, Frenohypon, Frenon, Frenormon, H 365, Hypophenon, Hypostat, Ibiopopp, Mepal, NSC 2834, NSC 33949, POP, Paroxon, Paroxypropion, Paroxypropione, Possipion, Possipione, Profenone, Proxiphenon, p-Hydroxyphenyl-1-propanone, p-Hydroxypropiophenone, p-Propionylphenol, p-Propiophenol
InChI Formula InChI=1S/C9H10O2/c1-2-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
SMILES CCC(=O)c1ccc(O)cc1
IUPAC Name 1-(4-hydroxyphenyl)propan-1-one
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.