missing translation for 'onlineSavingsMsg'
Learn More

p130 Antibody (2C8A8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 552.00€

Specifications

Antigen p130
Clone 2C8A8
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18200830
View Documents
Novus Biologicals
NBP2-52551
0.1 mg
552.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18603119
View Documents
Novus Biologicals
NBP2-52551-0.025mg
0.025 mg
203.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p130 Monoclonal specifically detects p130 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

p130
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF
Unconjugated
Mouse
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
PBS with 0.05% Sodium Azide
5934
Purified recombinant fragment of human RBL2 (AA: 939-1139) expressed in E. Coli.
Primary
128.4 kDa
2C8A8
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Purified
RUO
Human
FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2
RBL2
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.