missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y10/P2RY10 Antibody (639329) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10158I
This item is not returnable.
View return policy
Description
P2Y10/P2RY10 Monoclonal antibody specifically detects P2Y10/P2RY10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| P2Y10/P2RY10 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 639329 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| G-protein coupled purinergic receptor P2Y10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10 | |
| NS0 mouse myeloma cell line transfected with human P2RY10, Accession # O00398 | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 27334 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction