missing translation for 'onlineSavingsMsg'
Learn More

p57Kip2 Mouse anti-Human, Mouse, Rat, Clone: 1A12H3, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen p57Kip2
Clone 1A12H3
Concentration 0.7 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16868995
View Documents
Proteintech
66794-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
16858995
missing translation for 'viewDocuments'
Proteintech
66794-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications

Specifications

p57Kip2
0.7 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BWCR, BWS, CDKN1C, KIP2, p57, p57 Kip2, p57Kip2, WBS
CDKN1C
IgG1
Protein G
Antibody
1A12H3
Western Blot
Unconjugated
Mouse
Mouse, Rat, Human
P49918, P49919
1028, 12577, 246060
p57Kip2 Fusion Protein Ag19927
Primary
-20°C
CDKN1C
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.