Learn More
P62 SQSTM1 Rabbit anti-Human, Unconjugated, Clone: 1D17, Proteintech
Rabbit Recombinant Antibody
149.00€ - 439.00€
Spezifikation
| Antigen | P62 SQSTM1 |
|---|---|
| Clone | 1D17 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Beschreibung
The P62, SQSTM1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human P62, SQSTM1. This antibody recognizes human antigen. The P62, SQSTM1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| P62 SQSTM1 | |
| Western Blot | |
| Unconjugated | |
| Human | |
| IgG |
| 1D17 | |
| Monoclonal | |
| Rabbit | |
| 8878 | |
| Primary |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.