missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (CL3748) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05501MFV450
This item is not returnable.
View return policy
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| p63/TP73L | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| amplified in squamous cell carcinoma, Chronic ulcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| This antibody was developed using a recombinant protein derived from Q9H3D4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 8626 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3748 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction