missing translation for 'onlineSavingsMsg'
Learn More

p70 S6 Kinase beta/S6K2 Rabbit anti-Human, Mouse, Rat, Clone: 5D8F5, Novus Biologicals™

Rabbit Monoclonal Antibody

190.00€ - 496.00€

Specifications

Antigen p70 S6 Kinase beta/S6K2
Clone 5D8F5
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18309426
View Documents
Novus Biologicals
NBP3-16752-20UL
20 μg
190.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18313277
View Documents
Novus Biologicals
NBP3-16752-100UL
100 μg
496.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p70 S6 Kinase beta/S6K2 Monoclonal antibody specifically detects p70 S6 Kinase beta/S6K2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

p70 S6 Kinase beta/S6K2
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase
Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human p70 S6 Kinase beta/S6K2 (Q9UBS0).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
5D8F5
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Autophagy, Cancer, Cell Cycle and Replication
PBS, 0.05% BSA, 50% glycerol, pH7.3
6199
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.