missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP2 Monoclonal Antibody (PAC-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528523
This item is not returnable.
View return policy
Description
This antibody recognizes an extracellular activation-induced conformational epitope PAC-1 on CD41/CD61 complex (gpIIb/IIIa), also known as integrin alpha IIb beta 3, a receptor which mediates platelet aggregation.
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1 and ERK2, is predominantly expressed in hematopoietic tissues, and is localized in the nucleus.
Specifications
| DUSP2 | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 15mM sodium azide; pH 8 | |
| Q05923 | |
| DUSP2 | |
| Human platelets. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| PAC-1 | |
| Unconjugated | |
| DUSP2 | |
| dual specificity phosphatase 2; dual specificity protein phosphatase 2; Dual specificity protein phosphatase PAC-1; dual-specificity phosphatase 2; Dusp2; PAC1; Pac-1; serine/threonine specific protein phosphatase | |
| Mouse | |
| Sequential chromatography | |
| RUO | |
| 1844 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction