missing translation for 'onlineSavingsMsg'
Learn More

Palladium Rod, 4.0mm (0.16 in.) dia., 99.95% (Metals basis), Thermo Scientific™

Product Code. 11307088
Change view
Click to view available options
Quantity:
25 mm
Unit Size:
25mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307088 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11307088 Supplier Thermo Scientific Alfa Aesar Supplier No. 014763.DH

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Form: Approx.1.5g/cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5-3-7440
Appearance Silver gray rod
Quantity 25 mm
Odor Odorless
Assay Percent Range 99.95% (Metals basis)
Form Rod
Molecular Formula Pd
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES [Pd]
IUPAC Name palladium
Molecular Weight (g/mol) 106.42
PubChem CID 23938
ChEBI CHEBI:33363
Chemical Name or Material Palladium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.