missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 1 Antibody Cocktail
GREENER_CHOICE

Antibody Cocktail

Brand:  Invitrogen™ MA549309

Product Code. 17911581

  • 415.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: human NCL-H460 whole cellhuman Hela whole cellhuman A431 whole cell. IHC: Human Placenta tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 1
Cocktail
100 μg/mL
Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
P04264, Q6IMF3
KRT1
mixture of several monoclonal cytokeratin clones.
RUO
300250, 3848
-20°C
Lyophilized
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2
Unconjugated
KRT1
67 kDa cytokeratin; CK1; CK-1; cytokeratin 1; cytokeratin-1; EHK; EHK1; epidermolytic hyperkeratosis 1; epithelial keratin 1; Epithelial keratin-1; EPPK; Hair alpha protein; K1; Kb1; KER1; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 86; keratin complex 2, basic, gene 1; keratin, type II cytoskeletal 1; Keratin-1; KRT1; krt1 {ECO:0000250; KRT1A; Krt-2.1; Krt2-1; Krt86; KRTA; NEPPK; type II keratin Kb1; type-II keratin Kb1; UniProtKB:P04264}
Mouse
100 μg
Primary
Human, Rat
Antibody
IgG1, IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.