Learn More
pan-keratin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26411-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a pan-keratin antibody.
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
| pan-keratin | |
| Polyclonal | |
| Unconjugated | |
| KRT10 | |
| 58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3861, 3866, 3868, 3880 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02533, P02538, P04264, P05787, P08727, P08729, P08779, P12035, P13645, P13647, P19012, P19013, P35908 | |
| KRT1, KRT10, KRT14, KRT15, KRT16, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8 | |
| KRT5 Fusion Protein Ag24184 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |