missing translation for 'onlineSavingsMsg'
Learn More

Para Red, TRC

Product Code. 30004587
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004587 10 g 10g
17900465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004587 Supplier Toronto Research Chemicals Supplier No. TRCP19197510G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6410-10-2
Molecular Formula C16 H11 N3 O3
Molecular Weight (g/mol) 293.28
Synonym 2-Naphthalenol, 1-[2-(4-nitrophenyl)diazenyl]- (ACI), 1-[2-(4-Nitrophenyl)diazenyl]-2-naphthalenol (ACI), 2-Naphthalenol, 1-[(4-nitrophenyl)azo]- (9CI), 2-Naphthol, 1-(p-nitrophenylazo)- (6CI), C.I. Pigment Red 1 (7CI, 8CI), 1-(2-Hydroxynaphthyl)-4'-nitroazobenzene, 1-[(4-Nitrophenyl)azo]-2-naphthol, 1-[(p-Nitrophenyl)azo]-2-naphthol, C.I. 12070, C.P. Britelite Toner RP 9, C.P. Para Toner Light RP 2, Carnelio Para Red BS, Carnelio Para Red YS, Conc. Para Red, Conc. Para Red YS, Dainichi Para Red Toner, Duplex Para Red XD 20-2900, Eljon Para Red, Federal Red, Isol Para Red B, Kromon Para Red BS, Kromon Para Red YS, Label Red, Lacal Red GN, Lake Red 2R, Lake Red PN, Lutetia Red B, Lutetia Red N, Monolite Fast Red B, Monolite Fast Red BA, No. 7 Conc. Deep Red, NSC 210792, NSC 70812, p-Nitraniline red, Para Nitraniline Red, Para Red, Para Red B 20-2080, Para Red BS, Para Red Dark, Para Red Dark RA 1280, Para Red Light, Para Red Light 10333, Para Red Light RA 1210, Para Red Toner, Para Red Toner RPT 52, Para Red YS, Para Toner, Para Toner B, Para Toner D, Para Toner Dark 5065, Para Toner Deep A 1757, Para Toner Light A 4329, Para Toner Light RT 370, Para Toner Red, Para Toner Y 20-1150, Pigment Red 1, Pigment Red B, Pigment Red G, Pigment Red R, Pigment Red Zh, Printofix Red HN, Recolite Para Red B, Recolite Para Red G, Sanyo Signal Red Pure, Siloton Red B, Siloton Red G, Symuler Fast Para Red, Tertropigment Red PAB, Tolyl Red D, Tolyl Red LL, Tolyl Red M
IUPAC Name 1-[(E)-(4-nitrophenyl)diazenyl]naphthalen-2-ol
SMILES Oc1ccc2ccccc2c1N=Nc3ccc(cc3)[N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H11N3O3/c20-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)19(21)22/h1-10,20H
Formula Weight 293.08
Chemical Name or Material para Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.