missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine hydrochloride, Dr. Ehrenstorfer

Product Code. 30327568
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327568 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327568 Supplier Dr. Ehrenstorfer Supplier No. DREC15893720
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paroxetine Hydrochloride
CAS 78246-49-8
Analyte or Component Names Paroxetine Hydrochloride
Shipping Condition +20°C
Molecular Formula C19 H20 F N O3 . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym Paroxetine hydrochloride anhydrous, (3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride anhydrous, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride (1:1), (3S,4R)-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3S,4R)- (9CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3S-trans)-, (-)-trans-4-(4-Fluorophenyl)-3-(3,4-methylenedioxyphenoxymethyl)piperidine hydrochloride, Aropax 20, BRL 29060 hydrochloride, BRL 29060A, Deroxat, Paroxet, Paroxetine hydrochloride, Paxil, Seroxat, Tagonis, Xanadol
InChI Formula InChI=1S/C19H20FNO3.ClH/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;/h1-6,9,14,17,21H,7-8,10-12H2;1H/t14-,17-;/m0./s1
SMILES Cl.Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrochloride
Molecular Weight (g/mol) 365.83
Quantity 100 mg
Formula Weight 365.1194 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.