missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine Hydrochloride, TRC

Product Code. 17990315
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990315 10 mg 10mg
17943625 50 mg 50mg
17973325 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990315 Supplier Toronto Research Chemicals Supplier No. TRCP20575010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78246-49-8
Molecular Formula C19 H20 F N O3 . Cl H
Molecular Weight (g/mol) 365.83
Synonym Paroxetine hydrochloride anhydrous, (3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride anhydrous, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride (1:1), (3S,4R)-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3S,4R)- (9CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3S-trans)-, (-)-trans-4-(4-Fluorophenyl)-3-(3,4-methylenedioxyphenoxymethyl)piperidine hydrochloride, Aropax 20, BRL 29060 hydrochloride, BRL 29060A, Deroxat, Paroxet, Paroxetine hydrochloride, Paxil, Seroxat, Tagonis, Xanadol
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrochloride
SMILES Cl.Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H20FNO3.ClH/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;/h1-6,9,14,17,21H,7-8,10-12H2;1H/t14-,17-;/m0./s1
Formula Weight 365.1194
Percent Purity >95%
Chemical Name or Material Paroxetine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.