missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Particulate Respirator K101, FFP1, Unvalved
Description
- Economic, disposable respirator
- Diamond-style front panel that holds its shape even in hot or humid conditions
- Flat-fold design for easy storage
- Colour-coded nose clip for easy identification
- Single-loop strap allows for easy adjustments for a comfortable fit
- Completely free from staples and comes individually wrapped
- Individually wrapped
- Meets clogging requirements as per EN 149:2001+A1:2009, ensuring prolonged respirator life while maintaining ease of breathing
Specifications
Specifications
| Certifications/Compliance | CE |
| Disposable | Single-use |
| Product Type | Fold-Flat Dust/Mist Respirator FFP1 |
| For Use With (Equipment) | Particulate Respirator |
| Includes | Unvalved |
| Model | 3M K101 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?