missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody (CL15073) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
Specifications
| Antigen | Parvalbumin |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL15073 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:5000-1:10000 |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?