missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ Jurkat NFAT Pathway Reporter Cell Line
Description
The PathHunter™ Jurkat NFAT Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the NFAT pathway. This cell line enables assessment of NFAT pathway stimulation based on changes in the expression of a NFAT response element-driven reporter protein. This product uses EFC technology. The included cell line expresses PL-tagged reporter protein under the control of a NFAT pathway-inducible transcriptional response element. Pathway activation induces expression of the PL-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | NFAT pathway stimulation |
| Cell Line | Jurkat |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Anti-CD3 antibody [OKT3] |
| Quantity | 2 Vials |
| Target Entry | NFAT |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?