missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax2 Antibody (PAX2/8671R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20755IR
583.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Pax2 Monoclonal antibody specifically detects Pax2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Pax2 | |
Monoclonal | |
DyLight 755 | |
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
Recombinant fragment (around aa 223-354) of human Pax2 protein (exact sequence is proprietary) | |
0.1 mL | |
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
5076 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
PAX2/8671R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction