Learn More
PAX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 51036-2-AP-150UL
 View more versions of this product
Product Code. 16876305
  • 410.00€ / 150µL
Description
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
Specifications
Specifications
PAX3
Polyclonal
Unconjugated
PAX3
CDHS, HUP2, paired box 3, Paired box protein Pax 3, PAX3, WS1
Rabbit
Antigen Affinity Chromatography
RUO
18505, 5077
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23760, P24610
PAX3
PAX3 Fusion Protein Ag0477
150 μL
Primary
Human, Mouse