missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax5/BSAP Antibody (CL14548) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26864AF488
This item is not returnable.
View return policy
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Pax5/BSAP | |
| Monoclonal | |
| Alexa Fluor 488 | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| This antibody was developed using a synthetic peptide derived from Q02548, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
| 5079 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| CL14548 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction