Learn More
Pax5/BSAP Antibody (CL14548) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26864C
Product Code. 30289951
  • 770.00€ / 0.10mL
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pax5/BSAP
Monoclonal
DyLight 650
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
This antibody was developed using a synthetic peptide derived from Q02548, with the exact immunogen sequence remaining proprietary.
0.1 mL
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
5079
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot
CL14548
50mM Sodium Borate
Mouse
Protein A purified