missing translation for 'onlineSavingsMsg'
Learn More

PBB Mix 5 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer

Product Code. 30313871
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313871 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313871 Supplier Dr. Ehrenstorfer Supplier No. DRELA21030500CY
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 59080-40-9, 59080-39-6, 59080-37-4, 60044-25-9, 59080-34-1, 13029-09-9
Quantity 1 mL
Concentration 2,2',4,4',5,5'-Hexabromobiphenyl 10 μg/mL∣2,2',4,5',6-Pentabromobiphenyl 10 μg/mL∣2,2',5,5'-Tetrabromobiphenyl 10 μg/mL∣2,2',5,6'-Tetrabromobiphenyl 10 μg/mL∣2,2',5-Tribromobiphenyl 10 μg/mL∣2,2'-Dibromobiphenyl 10 μg/mL∣2,3',5-Tribromobiphenyl 10 μg/mL∣2,4',5-Tribromobiphenyl 10 μg/mL∣2,4,6-Tribromobiphenyl 10 μg/mL∣2,4-Dibromobiphenyl 10 μg/mL∣2,5-Dibromobiphenyl 10 μg/mL∣2,6-Dibromobiphenyl 10 μg/mL∣3,3',5,5'-Tetrabromobiphenyl 10 μg/mL∣3,4,5-Tribromobiphenyl 10 μg/mL∣4,4'-Dibromobiphenyl 10 μg/mL∣Decabromobiphenyl 10 μg/mL∣PBB-No. 1 10 μg/mL∣PBB-No. 155 10 μg/mL∣PBB-No. 2 10 μg/mL∣PBB-No. 29 10 μg/mL∣PBB-No. 3 10 μg/mL∣PBB-No.101 10 μg/mL∣PBB-No.49 10 μg/mL
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.