missing translation for 'onlineSavingsMsg'
Learn More

PBDE 15, TRC

Product Code. 30137023
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137023 25 g 25g
30134654 5 g 5g
17951375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137023 Supplier Toronto Research Chemicals Supplier No. TRCP21518525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-47-7
Molecular Formula C12 H8 Br2 O
Molecular Weight (g/mol) 328
Synonym Benzene, 1,1'-oxybis[4-bromo-, Ether, bis(p-bromophenyl) (6CI,7CI,8CI), 1,1'-Oxybis[4-bromobenzene], 4,4'-Dibromodiphenyl ether, BDE 15, Bis(4-bromophenyl) ether, Bis(bromophenyl) ether, Bis(p-bromophenyl) ether, Di(4-bromophenyl) ether, NSC 1787, NSC 9487, PBDE 15, p,p'-Dibromodiphenyl ether
IUPAC Name 1-bromo-4-(4-bromophenoxy)benzene
SMILES Brc1ccc(Oc2ccc(Br)cc2)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H8Br2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H
Formula Weight 325.8942
Chemical Name or Material PBDE 15 (4,4'-Dibromodiphenyl Ether)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.