missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBEF/Visfatin/NAMPT Antibody (362616), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4044A
This item is not returnable.
View return policy
Description
PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| PBEF/Visfatin/NAMPT | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 362616 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
| Mouse myeloma cell line NS0-derived recombinant mouse PBEF/Visfatin, Met1-His491, Accession # Q99KQ4 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity | |
| 10135 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction