Learn More
Melford PBS, 100 ml Tablets
Product Code. 30018687
Change view
Click to view available options
Quantity:
100 Tablets
Unit Size:
100 tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018687 100 Tablets 100 tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018687 Supplier Melford Supplier No. P32080100T
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PBS, 100 ml Tablets refer to pre-weighed tablets that are used to prepare a specific volume of PBS (Phosphate Buffered Saline) solution.
PBS, 100 ml Tablets refer to pre-weighed tablets that are used to prepare a specific volume of PBS (Phosphate Buffered Saline) solution.
PBS is a widely used buffer solution in biological and biochemical research. PBS serves as a versatile buffer for various applications, including cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. It helps maintain a stable pH and osmolarity, providing optimal conditions for biological reactions and preserving the integrity of cells and biomolecules.
Each tablet contains pre-determined amounts of sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic. When dissolved in distilled water or a suitable solvent, the tablet dissolves, releasing the components, and instantly forms a ready-to-use PBS solution with a total volume of 100 ml.
These tablets ensure accurate and consistent preparation of PBS, eliminating the need for weighing and measuring individual components. They are designed to simplify the process and save time in laboratory workflows.
Specifications
Quantity 100 Tablets