Learn More
Thermo Scientific PC Thickwall Tube
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Centrifuges
Unit Size:
Pack of 10
Description
- PC Thickwall tube, 25 mL x 10, for micro-ultracentrifuge
Specifications
Specifications
| Type | Thickwall Tube |
| Material | Polycarbonate |
| For Use With (Equipment) | Centrifuges |
| Disposable | Reusable |
| Capacity | 10 x 25 mL |
| Description | PC Thickwall Tube, 25 mL x 10, for micro-ultracentrifuge |