Learn More
PCB Mixture for HJ 350-2007 100 μg/mL in Hexane, Dr. Ehrenstorfer
Product Code. 30316327
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316327 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30316327 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000587HE
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 16605-91-7, 2051-60-7, 37680-73-2, 38380-03-9, 35065-28-2, 52712-04-6
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane