Learn More
PCDH7 Monoclonal Antibody (OTI2B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505447
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes a protein with an extracellular domain containing 7 cadherin repeats. The gene product is an integral membrane protein that is thought to function in cell-cell recognition and adhesion. Alternative splicing yields isoforms with unique cytoplasmic tails.Specifications
| PCDH7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60245 | |
| PCDH7 | |
| Full length human recombit protein of human PCDH7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2B12 | |
| Unconjugated | |
| PCDH7 | |
| BH-Pcdh, BHPCDH, PPP1R120 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5099 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.