Learn More
PCDH7 Monoclonal Antibody (OTI2B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505447S
Description
PCDH7 Monoclonal antibody specifically detects PCDH7 in Human samples. It is validated for Western BlotSpecifications
| PCDH7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60245 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5099 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2B12 | |
| Unconjugated | |
| PCDH7 | |
| BHPCDH; BH-Pcdh; BH-protocadherin (brain-heart); Brain-heart protocadherin; cell adhesion molecule; LOW QUALITY PROTEIN: protocadherin-7; PCDH7; PPP1R120; protein phosphatase 1, regulatory subunit 120; protocadherin 42-like protein; protocadherin 7; protocadherin 7; BH-protocadherin; BH-protocadherin (brain-heart); protocadherin-7; protocadherin-7; LOW QUALITY PROTEIN: protocadherin-7 | |
| Full length human recombinant protein of human PCDH7 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.