missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCPE-1/PCOLCE Antibody (CL6567) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76506CL1
This item is not returnable.
View return policy
Description
PCPE-1/PCOLCE Monoclonal antibody specifically detects PCPE-1/PCOLCE in Human samples. It is validated for Western Blot
Specifications
| PCPE-1/PCOLCE | |
| Monoclonal | |
| CoraFluor 1 | |
| PCPE, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein | |
| This antibody was developed using a recombinant protein derived from Q15113, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 5118 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| CL6567 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction