missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14827-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the formation of CDP-ethanolamine from CTP and phosphoethanolamine in the Kennedy pathway of phospholipid synthesis. Alternative splicing results in multiple transcript variants.Specifications
| PCYT2 | |
| Polyclonal | |
| Unconjugated | |
| PCYT2 | |
| ET, PCYT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5833, 68671, 89841 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88637, Q922E4, Q99447 | |
| PCYT2 | |
| PCYT2 Fusion Protein Ag6616 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction