missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (J116) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-43107-0.025mg
This item is not returnable.
View return policy
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| PD-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot | |
| J116 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| PDCD1 | |
| The immunogen for this antibody was PD1. | |
| 0.025 mg | |
| Apoptosis, Stem Cell Markers | |
| 5133 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction