missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (PDCD1/8170R), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20965AF488
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| PD-1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Recombinant fragment (around aa1-200) of human PD-1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| PDCD1/8170R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction