missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1/TGF beta Biosimilar Recombinant Human Monoclonal Antibody

PD-L1/TGF beta Biosimilar Recombinant Monoclonal Antibody
Brand: Invitrogen MA560149
This item is not returnable.
View return policy
Description
PD-L1/TGF beta Biosimilar Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Specifications
| PD-L1/TGF beta Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction