Learn More
PDCD1 Monoclonal Antibody (OTI9G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808034S
Description
PDCD1 Monoclonal antibody specifically detects PDCD1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| PDCD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q02242, Q15116 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 18566, 5133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry | |
| OTI9G6 | |
| Unconjugated | |
| Pdcd1 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| Full length human recombinant protein of human PDCD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.