missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD10 Mouse anti-Human, Clone: 1E5A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66440-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes an evolutionarily conserved protein associated with cell apoptosis. The protein interacts with the serine/threonine protein kinase MST4 to modulate the extracellular signal-regulated kinase (ERK) pathway. It also interacts with and is phosphoryated by serine/threonine kinase 25, and is thought to function in a signaling pathway essential for vascular developent. Mutations in this gene are one cause of cerebral cavernous malformations, which are vascular malformations that cause seizures and cerebral hemorrhages. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| PDCD10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BUL8 | |
| PDCD10 | |
| PDCD10 Fusion Protein Ag0348 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1E5A8 | |
| Unconjugated | |
| PDCD10 | |
| CCM3, PDCD10, programmed cell death 10, TFAR15 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11235 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction