missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17407-1-AP-150UL
This item is not returnable.
View return policy
Description
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL2 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. PDCL2 has been shown to interact with the chaperonin CCT and is essential for cytoskeletal protein function and cell cycle progression.Specifications
| PDCL2 | |
| Polyclonal | |
| Unconjugated | |
| PDCL2 | |
| GCPHLP, PDCL2, phosducin like 2, Phosducin like protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132954, 498352, 79455 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q78Y63, Q8N4E4 | |
| Pdcl2 | |
| PDCL2 Fusion Protein Ag11427 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction