Learn More
PDCL3 Monoclonal Antibody (OTI8D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809677
Description
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.Specifications
| PDCL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H2J4 | |
| PDCL3 | |
| Full length human recombit protein of human PDCL3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI8D6 | |
| Unconjugated | |
| PDCL3 | |
| HTPHLP, PHLP2A, PHLP3, VIAF, VIAF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79031 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.