missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38546-100ul
This item is not returnable.
View return policy
Description
PDE3B Polyclonal antibody specifically detects PDE3B in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PDE3B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CGIP1, CGI-PDE B, CGIPDE1, cGMP-inhibited 3'-5'-cyclic phosphodiesterase B, Cyclic GMP-inhibited phosphodiesterase B, cyclic nucleotide phosphodiesterase, EC 3.1.4, EC 3.1.4.17, HcGIP1, phosphodiesterase 3B, cGMP-inhibited | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-660 of human PDE3B (NP_000913.2).,, Sequence:, PLTPFPGFYPCSEIEDPAEKGDRKLNKGLNRNSLPTPQLRRSSGTSGLLPVEQSSRWDRNNGKRPHQEFGISSQGCYLNGPFNSNLLTIPKQRSSSVSLTHHVGLRRAGVLSSLSPVNSSNHGPVSTGSLTNRSPIEFPDTADFLNKPSVILQRSLGNAPNTPDFYQQLRNSDSNLCNSCGHQMLKYVSTSESDGTDCCSGKSGEEENIFSKESFKLMETQQEEETEKKDSRKLFQEGDKWLTEEAQSEQQTNIEQEVSLD | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5140 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction