Learn More
PDE4A Monoclonal Antibody (OTI5C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501206
Description
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.Specifications
| PDE4A | |
| Monoclonal | |
| 0.96 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P27815 | |
| PDE4A | |
| Full length human recombit protein of human PDE4A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C6 | |
| Unconjugated | |
| PDE4A | |
| DPDE2, PDE4, PDE46 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5141 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.