Learn More
PDGF R beta Antibody (PR7212), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1263PECY7
Product Code. 30028059
  • 863.00€ / 0.25mL
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGF R beta
Monoclonal
PE-Cyanine7
PBS
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Flow Cytometry
PR7212
Flow Cytometry
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Human skin fibroblast membrane extracts
0.25 mL
Oncogenes, Phospho Specific, Protein Kinase
5159
Store at 4C in the dark. Do not freeze.