missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDHA1/PDHA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517190
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The pyruvate dehydrogenase complex catalyzes the overall conversion of pyruvate to acetyl-CoA and CO2, and thereby links the glycolytic pathway to the tricarboxylic cycle.
Specifications
| PDHA1/PDHA2 | |
| Polyclonal | |
| Unconjugated | |
| PDHA2 | |
| PDHA; PDHA1; Pdha-1; PDHA2; Pdha-2; PDHAD; Pdhal; PDHCE1A; PDHE1-A type I; PDHE1-A type II; PHE1A; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 2; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; Pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1alpha subunit; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117098, 18597, 18598, 29554, 5160, 5161 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 56 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P08559, P26284, P29803, P35486, P35487, Q06437 | |
| PDHA1, PDHA2 | |
| Synthetic peptide corresponding to the sequences of both a1 and a2 subunits of human pyruvate dehydrogenase. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction