Learn More
PDPN Monoclonal Antibody (OTI7B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811746S
Description
PDPN Monoclonal antibody specifically detects PDPN in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PDPN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86YM7, Q9Z214, Q9Z2Y3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26556, 29546, 9456 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7B4 | |
| Unconjugated | |
| Homer1 | |
| 29kDa cytosolic podoplanin intracellular domain; Aggrus; CTA-520D8.1; E11; E11 antigen epitope; epithelial cell surface transmembrane protein antigen; glycoprotein 36; Glycoprotein 38; glycoprotein, 36-KD; GP36; Gp38; GP40; HT1A-1; hT1alpha-1; hT1alpha-2; lung type I cell membrane associated glycoprotein; lung type-I cell membrane-associated glycoprotein (T1A-2); Ots8; OTS-8; OTTHUMP00000009640; OTTHUMP00000044504; PA2.26; PA2.26 antigen; PDPN; PICD; Podoplanin; PSEC0003; PSEC0025; pulmonary type I alveolar epithelial cell transmembrane differentiation marker; RANDAM-2; RP23-348F1.2; RTI140; RTI40; T1a; T1A2; T1A-2; T1alpha; T1-alpha; TI1A; Transmembrane glycoprotein E11; type I cell 40 kDa protein | |
| Human recombinant protein fragment corresponding to amino acids 1-354 of human HOMER1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.