missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Mouse anti-Human, Clone: PCRP-PDX1-2C11, Novus Biologicals™
Mouse Monoclonal Antibody
508.00€
Specifications
| Antigen | PDX-1/IPF1 |
|---|---|
| Clone | PCRP-PDX1-2C11 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| PDX-1/IPF1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3651 | |
| IgG2c | |
| Protein A or G purified |
| PCRP-PDX1-2C11 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| Recombinant full-length human PDX-1/IPF1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title