missing translation for 'onlineSavingsMsg'
Learn More

Peanut Oil, TRC

Product Code. 30426548
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426548 10 mL 10mL
30423491 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426548 Supplier Toronto Research Chemicals Supplier No. TRCP80020010ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-03-7
Molecular Formula C30H45N9O5
Molecular Weight (g/mol) 611.7
Synonym Peanut oil or Groundnut oil (05C, 06C, 07C), Arachis hypogaea hypogaea seed oil, Arachis oil, Earthnut oil, Fats and Glyceridic oils, arachis, Fats and Glyceridic oils, groundnut, Fats and Glyceridic oils, peanut, Groundnut oil, Katchung oil, Peanut oils
SMILES C1=CC=C(C=C1)CC(C(=O)NC(CCCCN)C(=O)N)NC(=O)C(CCCN=C(N)N)NC(=O)C(CC2=CC=C(C=C2)O)N

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C30H45N9O5/c31-15-5-4-9-23(26(33)41)37-29(44)25(18-19-7-2-1-3-8-19)39-28(43)24(10-6-16-36-30(34)35)38-27(42)22(32)17-20-11-13-21(40)14-12-20/h1-3,7-8,11-14,22-25,40H,4-6,9-10,15-18,31-32H2,(H2,33,41)(H,37,44)(H,38,42)(H,39,43)(H4,34,35,36)
Chemical Name or Material Peanut Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.