missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEAR1 Antibody (492621) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4527JF669
This item is not returnable.
View return policy
Description
PEAR1 Monoclonal antibody specifically detects PEAR1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| PEAR1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| JEDI, platelet endothelial aggregation receptor 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human PEAR1, Leu21-Ser754, Accession # Q5VY43 | |
| RUO | |
| 375033 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF | |
| 492621 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction